missing translation for 'onlineSavingsMsg'
Learn More

Vinyl Laurate (stabilized with MEHQ) 99.0+%, TCI America™

Catalog No. L009025ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L009025ML 25 mL
L0090500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. L009025ML Supplier TCI America Supplier No. L009025ML

Chemical Identifiers

CAS 2146-71-6
Molecular Formula C14H26O2
Molecular Weight (g/mol) 226.36
MDL Number MFCD00048421
InChI Key GLVVKKSPKXTQRB-UHFFFAOYSA-N
Synonym vinyl laurate, dodecanoic acid, ethenyl ester, vinyl dodecanoate, lauric acid, vinyl ester, lauric acid vinyl ester, lauric acid vinyl, acmc-209t6q, dodecanoic acid,ethenyl ester, lauric acid, vinyl ester 8ci, vinyl laurate gc
PubChem CID 75069
IUPAC Name ethenyl dodecanoate
SMILES CCCCCCCCCCCC(=O)OC=C

Specifications

Color Yellow
Boiling Point 285°C
Quantity 25 mL
Formula Weight 226.36
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Vinyl Laurate (stabilized with MEHQ)
EINECSNumber (2)-3309
TSCA Yes