missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Vinyl ESD Chair - Medium Bench Height with Seat Tilt and ESD Casters in ESD Vinyl

Fisherbrand™ Vinyl ESD Chairs are height adjustable for 36 in. work surfaces & feature ergonomic adjustments, electro-static components such as Vinyl & self-braking casters, & adjustable arms. Chairs are manufactured to ensure proper grounding for static sensitive applications.

$1782.80 - $1782.80

Specifications

Adjustable Yes
Adjustable (Height) 21.5 to 29 in.
Autoclavable Not Autoclavable
Back Style Standard
Base Material Polished Cast Aluminum
View More Specs

Products 1
Catalog Number Mfr. No. Color Price Quantity  
Catalog Number Mfr. No. Color Price Quantity  
14-400-282
View Documents
Fisherbrand™
VESD27BLKT
Black
Each for $1,782.80
Only null left
 
Description

Description

  • Adjustable from 21.5-29 inches from the top of the seat to the floor
  • Features ergonomic adjustments including seat height
  • Locking back tilt
  • Locking seat tilt
  • Electro-static dissipating components to ensure proper grounding
  • Cast-aluminum base
  • Dual-wheel, self-braking, non-marring ESD casters suitable for hard floors
  • Electro-static dissipating vinyl
  • Meets ANSI/ESD STM12.1-2013
Specifications

Specifications

Yes
Not Autoclavable
Polished Cast Aluminum
Electro-Static Dissipating, dual-wheel, non-marring casters (suitable for both hard and carpeted floors)
136 kg
Polyurethane foam, heavy-duty ESD fabric
33 lb.
Yes
No
Yes
Vinyl
21.5 to 29 in.
Standard
ANSI/ESD-STM-12.1-2006
300 lb.
Medium Bench Height
Ergonomic 8 degree forward and 3 degree backward seat tilt locks in place
No
Yes
Yes
ESD Chair
Medium Bench Height
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'