missing translation for 'onlineSavingsMsg'
Learn More

Vinyl Decanoate (stabilized with MEHQ) 99.0+%, TCI America™

Catalog No. C089525ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C089525ML 25 mL
C0895500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C089525ML Supplier TCI America Supplier No. C089525ML

Chemical Identifiers

CAS 4704-31-8
Molecular Formula C12H22O2
Molecular Weight (g/mol) 198.306
MDL Number MFCD00027349
InChI Key CMDXMIHZUJPRHG-UHFFFAOYSA-N
Synonym vinyl decanoate, decanoic acid, vinyl ester, vinyl n-decanoate, unii-ff00i399li, decanoic acid, ethenyl ester, n-capric acid vinyl ester, decanoic acid vinyl, acmc-1aqz4, decanoic acid vinyl ester, capric acid vinyl ester = vinyl decenoate
PubChem CID 62140
IUPAC Name ethenyl decanoate
SMILES CCCCCCCCCC(=O)OC=C

Specifications

Color Colorless
Boiling Point 148°C
Quantity 25 mL
Formula Weight 198.31
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Vinyl Decanoate (stabilized with MEHQ)
EINECSNumber (2)-0761
RTECSNumber HE0527500
TSCA No