missing translation for 'onlineSavingsMsg'
Learn More

Vinyl Crotonate (stabilized with MEHQ) 99.0+%, TCI America™

Catalog No. p-7124672
Change view
Click to view available options
Quantity:
25 mL
500 mL
Catalog No. Quantity
C089725ML 25 mL
C0897500ML 500 mL
2 options

Catalog No. C089725ML

Supplier: TCI America C089725ML

Only null left

Chemical Identifiers

CAS 14861-06-4
Molecular Formula C6H8O2
Molecular Weight (g/mol) 112.128
MDL Number MFCD00009288
InChI Key IYNRVIKPUTZSOR-HWKANZROSA-N
Synonym vinyl crotonate, vinyl 2-butenoate, crotonic acid, vinyl ester, crotonic acid vinyl ester, unii-smc9a01nvs, ethenyl e-but-2-enoate, vinylester kyseliny krotonove, vinyl but-2-enoate, smc9a01nvs, vinylester kyseliny krotonove czech
PubChem CID 5365800
IUPAC Name ethenyl (E)-but-2-enoate
SMILES CC=CC(=O)OC=C

Specifications

Melting Point -60°C
Color Colorless
Boiling Point 134°C
Quantity 25 mL
UN Number 3272
Formula Weight 112.13
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Vinyl Crotonate (stabilized with MEHQ)
EINECSNumber (2)-4046
RTECSNumber GQ5850000
TSCA No
Recommended Storage Refrigerator