accessibility menu, dialog, popup

UserName

CAS RN 14861-06-4

CH3OOCH2(Z)

CAS RN 14861-06-4

IUPAC Name: ethenyl (2Z)-but-2-enoate
Synonyms: ethenyl (2Z)-but-2-enoate
Molecular Weight (g/mol): 112.13
Molecular Formula: C6H8O2
InChi Key: IYNRVIKPUTZSOR-HYXAFXHYSA-N
SMILES: C\C=C/C(=O)OC=C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results