missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vinyl Crotonate (stabilized with MEHQ) 99.0+%, TCI America™
Specifications
Specifications
| Melting Point | -60°C |
| Color | Colorless |
| Boiling Point | 134°C |
| Quantity | 25 mL |
| UN Number | 3272 |
| Formula Weight | 112.13 |
| Percent Purity | ≥99.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Vinyl Crotonate (stabilized with MEHQ) |
Safety and Handling
| EINECSNumber | (2)-4046 |
| RTECSNumber | GQ5850000 |
| TSCA | No |
| Recommended Storage | Refrigerator |