missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vinyl Cinnamate (stabilized with MEHQ) 99.0+%, TCI America™
$121.28 - $823.97
Chemical Identifiers
| CAS | 3098-92-8 |
|---|---|
| Molecular Formula | C11H10O2 |
| Molecular Weight (g/mol) | 174.199 |
| MDL Number | MFCD00059324 |
| InChI Key | WGXGKXTZIQFQFO-CMDGGOBGSA-N |
| Synonym | Cinnamic Acid Vinyl Ester |
| PubChem CID | 5371819 |
| IUPAC Name | ethenyl (E)-3-phenylprop-2-enoate |
| SMILES | C=COC(=O)C=CC1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
C089925ML
|
TCI America
C089925ML |
25 mL |
Each for $121.28
|
|
|||||
|
C0899500ML
|
TCI America
C0899500ML |
500 mL |
Each for $823.97
|
|
|||||
Chemical Identifiers
| 3098-92-8 | |
| 174.199 | |
| WGXGKXTZIQFQFO-CMDGGOBGSA-N | |
| 5371819 | |
| C=COC(=O)C=CC1=CC=CC=C1 |
| C11H10O2 | |
| MFCD00059324 | |
| Cinnamic Acid Vinyl Ester | |
| ethenyl (E)-3-phenylprop-2-enoate |
Specifications
| 3098-92-8 | |
| Yellow | |
| C11H10O2 | |
| 25 mL | |
| WGXGKXTZIQFQFO-CMDGGOBGSA-N | |
| ethenyl (E)-3-phenylprop-2-enoate | |
| 5371819 | |
| ≥99.0% (GC) | |
| Vinyl Cinnamate (stabilized with MEHQ) |
| -28°C | |
| 125°C | |
| MFCD00059324 | |
| Cinnamic Acid Vinyl Ester | |
| C=COC(=O)C=CC1=CC=CC=C1 | |
| 174.199 | |
| 174.20 | |
| Liquid |
Safety and Handling
EINECSNumber : (3)-4655
TSCA : No