missing translation for 'onlineSavingsMsg'
Learn More

Vinyl(chloromethyl)dimethylsilane, 97%

Catalog No. p-7044014
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0405606 5 g
AAL0405614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0405606 Supplier Thermo Scientific Chemicals Supplier No. L0405606

CAS: 16709-86-7 | C5H11ClSi | 134.678 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 16709-86-7
Molecular Formula C5H11ClSi
Molecular Weight (g/mol) 134.678
MDL Number MFCD00040860
InChI Key SZZZMXFBEKWPBU-UHFFFAOYSA-N
Synonym chloromethyl dimethylvinylsilane, silane, chloromethyl ethenyldimethyl, vinyl chloromethyl dimethylsilane, chloromethyl ethenyldimethylsilane, ccris 2448, chloromethyldimethylvinylsilane, silane, chloromethyl dimethylvinyl, chloromethyldimethylvinyl silane, chloromethyl dimethyl vinyl silane, chloromethyl ethenyl dimethylsilane
PubChem CID 85556
IUPAC Name chloromethyl-ethenyl-dimethylsilane
SMILES C[Si](C)(CCl)C=C

Specifications

Boiling Point 121°C to 122°C
Quantity 5 g
UN Number UN1993
Beilstein 2071374
Flash Point 14°C (57°F)
Formula Weight 134.68
Percent Purity 97%
Refractive Index 1.443
Density 0.893
Chemical Name or Material Vinyl(chloromethyl)dimethylsilane
Hazard Category H225-H315-H319-H335
Hazard Statement GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 240-758-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only