CAS RN 16709-86-7
IUPAC Name:
(chloromethyl)(ethenyl)dimethylsilane
Synonyms:
(chloromethyl)(ethenyl)dimethylsilane
Molecular Weight (g/mol):
134.68
Molecular Formula:
C5H11ClSi
InChi Key:
SZZZMXFBEKWPBU-UHFFFAOYSA-N
SMILES:
C[Si](C)(CCl)C=C
1
–
1
of
1
results
1
Promotions Available
Vinyl(chloromethyl)dimethylsilane, 97%
Vinyl(chloromethyl)dimethylsilane, 97%
CAS: 16709-86-7 Molecular Formula: C5H11ClSi Molecular Weight (g/mol): 134.678 MDL Number: MFCD00040860 InChI Key: SZZZMXFBEKWPBU-UHFFFAOYSA-N Synonym: chloromethyl dimethylvinylsilane,silane, chloromethyl ethenyldimethyl,vinyl chloromethyl dimethylsilane,chloromethyl ethenyldimethylsilane,ccris 2448,chloromethyldimethylvinylsilane,silane, chloromethyl dimethylvinyl,chloromethyldimethylvinyl silane,chloromethyl dimethyl vinyl silane,chloromethyl ethenyl dimethylsilane PubChem CID: 85556 IUPAC Name: chloromethyl-ethenyl-dimethylsilane SMILES: C[Si](C)(CCl)C=C