missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Vinyl Chair - Medium Bench Height with Round Tube Base, Adjustable Arms, and Casters in Grade B Vinyl

Fisherbrand™ Vinyl Chairs with Round Tube Base are height adjustable for 36 in. work surfaces and feature ergonomic adjustments, adjustable arms, fixed foot ring, and self-braking casters. Antibacterial/Antifungal Grade B vinyl in a variety of colors will suit any work space.

Supplier:  Fisherbrand™ 91262PBLKCAA_RTE

 View more versions of this product

Catalog No. 14-359-074


Only null left
Explore available promotions

Description

Description

  • Adjustable from 25.25-33 inches from the top of the seat to the floor
  • Features ergonomic adjustments including seat height
  • Locking back tilt
  • 6-way adjustable arm rests adjust up and down, in and out, and twist toward and away from the user
  • Black, powder-coated steel base with fixed foot ring
  • Dual-wheel, self-braking, non-marring casters suitable for hard floors
  • Antibacterial/antifungal Grade B Vinyl
  • Variety of color options to suit any work space
Specifications

Specifications

15 year parts, lifetime on pneumatic lift, 5 year on upholstery
No
19 in.
42 lb.
No
18 in.
Powder-coated Steel
2.5 in.
Yes
Black dual-wheel, self-braking, non-marring casters (suitable for both hard and carpeted floors)
Medium Bench Height
Yes
Vinyl
Powder-coated steel with fixed foot ring
25.25 to 33 in.
Yes
300 lb.
General Seating Chair
Yes
25 in.
Yes (Pneumatic)
6-way adjustable (move up and down, in and out, and twist toward or away from the user)
Standard
136 kg
Black
Medium Bench Height
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions