missing translation for 'onlineSavingsMsg'
Learn More

Vinyl 2-Chlorobenzoate (stabilized with HQ) 98.0+%, TCI America™

Catalog No. C10485G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C10485G 5 g
C104825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C10485G Supplier TCI America Supplier No. C10485G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 15721-27-4
Molecular Formula C9H7ClO2
Molecular Weight (g/mol) 182.603
MDL Number MFCD00059479
InChI Key ZJIHUSWGELHYBJ-UHFFFAOYSA-N
Synonym 2-Chlorobenzoic Acid Vinyl Ester
PubChem CID 587335
IUPAC Name ethenyl 2-chlorobenzoate
SMILES C=COC(=O)C1=CC=CC=C1Cl

Specifications

Color Yellow
Boiling Point 130°C
Quantity 5 g
Formula Weight 182.60
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Vinyl 2-Chlorobenzoate (stabilized with HQ)