accessibility menu, dialog, popup

UserName

H2COOCl

CAS RN 15721-27-4

IUPAC Name: ethenyl 2-chlorobenzoate
Synonyms: ethenyl 2-chlorobenzoate
Molecular Weight (g/mol): 182.6
Molecular Formula: C9H7ClO2
InChi Key: ZJIHUSWGELHYBJ-UHFFFAOYSA-N
SMILES: ClC1=CC=CC=C1C(=O)OC=C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results