missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VILL Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5120362
Description
Positive test controls include: Mouse kidney, Mouse eye, Mouse lung, Rat kidney, Rat eye. Immunogen sequence: LDKYDIMLLD TWQEIFLWLG EAASEWKEAV AWGQEYLKTH PAGRSPATPI VLVKQGHEPP TFIGWFFTWD PYKWTSHPSH KEVVDGSPAA ASTISEITAE VNNLRLSRWP GNGRAGAVAL QALKGSQDSS ENDLVRSPKS AGSRTSSSVS STSATINGGL RREQLMHQAV EDLPEGVDPA RREFYLSDSD FQDIFGKSKE EFYSMATWRQ RQEKKQLGFF.
The protein encoded by this gene belongs to the villin/gelsolin family. It contains 6 gelsolin-like repeats and a headpiece domain. It may play a role in actin-bundling. [provided by RefSeq].Specifications
| VILL | |
| Polyclonal | |
| Unconjugated | |
| VILL | |
| VILL; villin like; villin-like; villin-like protein | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 22351, 301057, 50853 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 2.78 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| O15195, Q91YD6 | |
| VILL | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 637-856 of human VILL (NP_056957.3). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |