missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vilazodone Hydrochloride 98.0+%, TCI America™
$395.17 - $1101.65
Chemical Identifiers
| CAS | 163521-08-2 |
|---|---|
| Molecular Formula | C26H28ClN5O2 |
| Molecular Weight (g/mol) | 477.993 |
| MDL Number | MFCD00940014 |
| InChI Key | RPZBRGFNBNQSOP-UHFFFAOYSA-N |
| Synonym | 5-[4-[4-(5-Cyano-1H-indol-3-yl)butyl]piperazin-1-yl]benzofuran-2-carboxamide Hydrochloride |
| PubChem CID | 6918313 |
| ChEBI | CHEBI:70705 |
| IUPAC Name | 5-[4-[4-(5-cyano-1H-indol-3-yl)butyl]piperazin-1-yl]-1-benzofuran-2-carboxamide;hydrochloride |
| SMILES | C1CN(CCN1CCCCC2=CNC3=C2C=C(C=C3)C#N)C4=CC5=C(C=C4)OC(=C5)C(=O)N.Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
V014950MG
|
TCI America
V014950MG |
50 mg |
Each for $395.17
|
|
|||||
|
V0149200MG
|
TCI America
V0149200MG |
200 mg |
Each for $1,101.65
|
|
|||||
Chemical Identifiers
| 163521-08-2 | |
| 477.993 | |
| RPZBRGFNBNQSOP-UHFFFAOYSA-N | |
| 6918313 | |
| 5-[4-[4-(5-cyano-1H-indol-3-yl)butyl]piperazin-1-yl]-1-benzofuran-2-carboxamide;hydrochloride |
| C26H28ClN5O2 | |
| MFCD00940014 | |
| 5-[4-[4-(5-Cyano-1H-indol-3-yl)butyl]piperazin-1-yl]benzofuran-2-carboxamide Hydrochloride | |
| CHEBI:70705 | |
| C1CN(CCN1CCCCC2=CNC3=C2C=C(C=C3)C#N)C4=CC5=C(C=C4)OC(=C5)C(=O)N.Cl |
Specifications
| 163521-08-2 | |
| White-Yellow | |
| MFCD00940014 | |
| 5-[4-[4-(5-Cyano-1H-indol-3-yl)butyl]piperazin-1-yl]benzofuran-2-carboxamide Hydrochloride | |
| C1CN(CCN1CCCCC2=CNC3=C2C=C(C=C3)C#N)C4=CC5=C(C=C4)OC(=C5)C(=O)N.Cl | |
| 477.993 | |
| CHEBI:70705 | |
| ≥98.0% (HPLC) | |
| Vilazodone Hydrochloride |
| 279°C | |
| C26H28ClN5O2 | |
| 50 mg | |
| RPZBRGFNBNQSOP-UHFFFAOYSA-N | |
| 5-[4-[4-(5-cyano-1H-indol-3-yl)butyl]piperazin-1-yl]-1-benzofuran-2-carboxamide;hydrochloride | |
| 6918313 | |
| 477.99 | |
| Crystalline Powder |
Safety and Handling
TSCA : No
Recommended Storage : Freezer (-20 °C)