Learn More
Vikan™ Waterfed Washing Brush with Angle Adjustment
Vikan™ Waterfed Washing Brush with Angle Adjustment is commonly used to clean walls and sensitive surfaces.
Supplier:  Vikan™ 70674
 View more versions of this product
Catalog No. 17227823
Only null left
Description
Description
  • Features split fiber bristles to absorb liquid
  • Allows the user to efficiently carry soapy water from bucket to the surface
  • Bristles are soft enough for sensitive surfaces
  • Waterfed design makes it excellent for rinsing after foaming
  • Neck angle is adjustable to allow all bristles to remain constantly in contact with the cleaning surface
  • Color-coded
Specifications
Chemical Identifiers
Cleaning Brush
Soft/Split
Polypropylene
FDA Compliant
9.25 in.
Medium
Polyester
Yes
Non-sterile
Thread type Euro Hygienic
1 Ea.
Specifications
Cleaning Brush
Soft/Split
Polypropylene
FDA Compliant
Thread type Euro Hygienic
1 Ea.
Polyester
Yes
Non-sterile
Red
9.25 in.
Medium
Product Suggestions
Product Suggestions