missing translation for 'onlineSavingsMsg'
Learn More

Vikan™ Stiff 12 in. Deck/Wall Scrub

Vikan™ Stiff 12 in. Deck/Wall Scrub can be used in conjunction with water and cleaning agents.

Supplier:  Vikan™ 70603

 View more versions of this product

Catalog No. 17-227-801

Only null left

Description

Description

  • Scrubbing broom with firm bristles
  • Bristle length is ideal for scrubbing all types of floors; epoxy, tiles, cement, etc
  • Reaches well into holes and grouting to remove hidden dirt
  • Color-coded
Specifications

Specifications

Scrub Brush
Stiff
Polypropylene
FDA Compliant
Thread type Euro Hygienic
1 Ea.
Polyester
Yes
Non-sterile
Blue
11.8 in.
Large
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions