missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vikan™ Spray Gun
Spray gun allows for a gentle flow or a high-pressured stream of water from an attached hose to clean different surfaces.
Supplier: Vikan™ 93243
Description
- Suitable for cleaning floors, work surfaces, walls, and machinery
- By applying a variable amount of tension on the rubber insulated trigger, the user can control the pressure
- The tool is suitable for many cleaning applications, including rinsing off work surfaces and walls after chemical foaming
- The water jet can also be adjusted using the screw at the end of gun
- An arrow on the water gun indicates the spray direction of the water jet
- Included adapter fits to our Automatic Shut Off Coupling (0703US) or the Hose Clamp (0701) can be used to attach it directly to a hose
Specifications
| Spray Gun | |
| Yes | |
| Medium | |
| 1 Ea. |
| Blue | |
| Rubber, Brass, Stainless Steel | |
| 5.7 x 7.1 in. | |
| 1.76 lb. |