Learn More
Vikan™ Medium Bench Brush
Vikan™ Medium Bench Brush is a dusting brush with a smooth, ergonomically designed handle.
Supplier: Vikan™ 45892
Description
- Smooth, ergonomically designed handle
- The medium strength bristles remove moist or medium sized particles, such as sugar, chocolate, pastry, vegetable peelings
- Can also be used as a dustpan brush on floors
- Color-coded
Specifications
| Bench Brush | |
| Yes | |
| Non-sterile | |
| Green | |
| 1 Ea. |
| Polyester | |
| Polypropylene | |
| FDA Compliant | |
| 13 in. | |
| Medium |