missing translation for 'onlineSavingsMsg'
Learn More

Vikan™ Ultra Hygiene Handle, 51 in.

Ultra hygiene handle is easy to clean and molded into a wider grip for comfort.

Supplier:  Vikan™ 29607

 View more versions of this product

Catalog No. 17227122


Only null left

Description

Description

  • Ergonomically designed grip for comfort during use
  • Made from food-grade, chemically resistant polypropylene
  • Hygienic design minimizes the risk of cross-contamination and makes it easy to clean and maintain
  • Color-coded
Specifications

Chemical Identifiers

Ultra Hygiene Handle
Non-sterile
1
FDA Compliant
Polypropylene
51 in.
1.25 in.
Thread type Euro Hygienic
Yes

Specifications

Ultra Hygiene Handle
51 in.
1.25 in.
Thread type Euro Hygienic
Yes
Orange
Non-sterile
1
FDA Compliant
Polypropylene
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions