Learn More
Vikan™ Single Blade Ultra Hygiene Bench Squeegee, 24 in.
Squeegee is ideal for removing water or food debris from food contact surfaces.
$67.14
Specifications
Product Type Single-blade Squeegee
Certifications/Compliance FDA Compliant
Description Thread type Euro Hygienic
Includes Thread type Euro Hygienic
Material Polypropylene, Rubber
View More Specs
Products 6
Catalog Number Mfr. No. Color Price Quantity & Availability  
Catalog Number Mfr. No. Color Price Quantity & Availability  
19830238
View Documents Promotion Details
Vikan™
71604
Red
Each for $67.14
Only null left
17227846
View Documents Promotion Details
Vikan™
71607
Orange
Each for $67.14
Only null left
19830132
View Documents Promotion Details
Vikan™
71606
Yellow
Each for $67.14
Only null left
17227847
View Documents Promotion Details
Vikan™
71608
Purple
Each for $67.14
Only null left
17227845
View Documents Promotion Details
Vikan™
71601
Pink
Each for $67.14
Only null left
19830237
View Documents Promotion Details
Vikan™
71605
White
Each for $67.14
Only null left
Description
Description
  • One-piece construction
  • Single-blade squeegee
  • Ultra-hygienic design that minimizes areas for water and food soils to collect
  • Effective in areas requiring the highest level of hygiene
  • The soft and flexible rubber blade is ideal for removing water or food debris from floors and food contact surfaces
  • Hygiene squeegees are a good foundation for any HACCP or food safety program
  • A splash guard helps prevent water from reaching the dried surface
  • Color-coded
Specifications
Specifications
Single-blade Squeegee
Thread type Euro Hygienic
Polypropylene, Rubber
Large
FDA Compliant
Thread type Euro Hygienic
1 Ea.
3.5 in.