Learn More
Vikan™ Condensation Squeegee, 16 in.
Condensation squeegee is intended to be used with a draining handle to collect condensation that forms in high areas such as overhead pipes and ceilings.
Supplier:  Vikan™ 77163
 View more versions of this product
Catalog No. 17227873
Only null left
Description
Description
  • Squeegee with drain holes
  • For effective removal of condensation from ceilings and pipes
  • Collection of condensation prevents moisture from dripping into food product or on costly machinery
  • Color-coded
Specifications
Chemical Identifiers
Floor Squeegee
Thread type Euro Hygienic
1 Ea.
4.7 in.
FDA Compliant
Polypropylene, TPE Rubber
Large
Specifications
Floor Squeegee
Blue
Thread type Euro Hygienic
1 Ea.
4.7 in.
FDA Compliant
Thread type Euro Hygienic
Polypropylene, TPE Rubber
Large
Product Suggestions
Product Suggestions