missing translation for 'onlineSavingsMsg'
Learn More

Vikan™ Extra Stiff Push Broom, 13 in.

Extra stiff push broom is ideal for sweeping large particles, such as meat and bones, in moist environments.

Supplier:  Vikan™ 29152

 View more versions of this product

Catalog No. 17227067


Only null left

Description

Description

  • Feature long, thick, and stiff bristles
  • Ideal for sweeping heavy debris
  • Suitable for use in wet and external areas
  • Color-coded
Specifications

Chemical Identifiers

Push Broom
Non-sterile
1
FDA Compliant
Polypropylene, Stainless Steel
13 in.
2.9 in.
Thread type Euro hygienic
Yes

Specifications

Green
13 in.
2.9 in.
Thread type Euro hygienic
Yes
Push Broom
Non-sterile
1
FDA Compliant
Polypropylene, Stainless Steel
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions