missing translation for 'onlineSavingsMsg'
Learn More

ViiA™ 7 Dx Instrument, 384 well, desktop

Catalog No. 4453541
Encompass
Change view
Click to view available options
Quantity:
1 instrument
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4453541 1 instrument
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 4453541 Supplier Thermo Fisher Scientific Supplier No. 4453541

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Block Format Interchangeable
Format 384-well Plate
Sensitivity Down to 1 Copy
Temperature Range (Metric) 4 to 100°C
Reaction Volume Range 5–20 μL
Dynamic Range Up to 9 Logs of Linear Dynamic Range
Content And Storage Desktop Computer
High-throughput Compatibility High-throughput Compatible, Multiplexing
For Use With (Equipment) ViiA™ 7 Dx System
Thermal Uniformity ±0.5°C
Reaction Speed Standard
Run Time About 35 min. (varies depending on Reagents and Other Factors)
Optics 6 Emission Filters (500 to 720 nm), 6 Excitation Filters (450 to 670 nm), OptiFlex™ System (Enhanced Fluorescence Detection)
Dimensions 53.3 cm (21 in.) (W) x 63.5 cm (25 in.) (D) x 64.5 cm (25.4 in.) (H)
Capacity 1 x 384-well Plate
Voltage 100 to 240 V
Electrical Requirements 60 Hz
Product Line ViiA
Display Type Touchscreen
Resolution Detects as little as 1.5-fold changes in target quantities (in singleplex reactions)
Quantity 1 instrument
Weight 60.7 kg
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.