missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Victoria Pure Blue BO, 0.03% (w/v), Ricca Chemical
Supplier: Ricca Chemical Company R91080004A
Spécifications
| Victoria Pure Blue BO | |
| 98.0,0.03 | |
| Natural Poly Bottle | |
| Blue |
| 2390-60-5,7732-18-5 | |
| 100.0,0.03 | |
| Aqueous Solution | |
| 4 L |