missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Vibrio parahaemolyticus serotype O3:K6 ompK Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5117599
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5117599 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5117599 Supplier Invitrogen™ Supplier No. PA5117599
Only null left

Rabbit Polyclonal Antibody

This antibody has been tested in direct-ELISA.

Serves as receptor for a broad-host-range vibriophage, KVP40.

Specifications

Antigen Vibrio parahaemolyticus serotype O3:K6 ompK
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene Alias Outer membrane protein OmpK
Host Species Rabbit
Immunogen Recombinant Vibrio parahaemolyticus serotype O3:K6 Outer membrane protein OmpK protein (21-266aa).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less