missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ VGLL4 Recombinant Rabbit Monoclonal Antibody (16H37L22)

Catalog No. 703699
Encompass
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
703699 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 703699 Supplier Invitrogen™ Supplier No. 703699
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rabbit Recombinant Monoclonal Antibody

This antibody is predicted to react with Mouse, Rat, Bovine.

Transcription cofactor vestigial-like protein 4 (VGLL4) acts as a specific coactivator for the mammalian TEFs.

Specifications

Antigen VGLL4
Applications Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 16H37L22
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2-7.4
Gene Vgll4
Gene Accession No. Q14135
Gene Alias BC048841; KIAA0121; Transcription cofactor vestigial-like protein 4; vestigial like 4; vestigial like 4 (Drosophila); vestigial like family member 4; Vestigial-like 4; vestigial-like family member 4; VGL-4; Vgll4
Gene Symbols Vgll4
Host Species Rabbit
Immunogen Peptides corresponding to human VGLL4 [1) aa6-aa23 2) aa235-aa253].
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9686
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less