missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VETRANAL™ Zilpaterol hydrochloride, MilliporeSigma™ Supelco™
Specifications
Specifications
| Product Line | VETRANAL |
| CAS | 119520-06-8 |
| Physical Form | Neat |
| Linear Formula | C14H19N3O2 · HCl |
| Synonym | (6 RS,7 RS)-4,5,6, 7-Tetrahydro-7-hydroxy-6-[(1-methylethyl)amino]imidazo[4,5,1-jk][1 ]benzazepin-2(1 H)-one hydrochloride |
| Type | Analytical Standard |
| Quantity | 10 mg |
| Formula Weight | 297.78 |
| Grade | Analytical Standard |
VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH