missing translation for 'onlineSavingsMsg'
Learn More

VETRANAL™ Vedaprofen, MilliporeSigma™ Supelco™

Catalog No. 111015322
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015322 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015322 Supplier Merck Emd Millipore Supplier No. 3253310MG

Vedaprofen is an arylpropionic acid derivative used as a non-steroidal anti-inflammatory drug (NSAID) in veterinary medicine.

Pharmaceutical (small molecule)

Specifications

Product Line VETRANAL
CAS 71109-09-6
Physical Form Neat
Linear Formula C19H22O2
MDL Number MFCD00866019
Synonym (+/-)-2-(4-Cyclohexylnaphthalen-1-yl)propionic acid
Type Analytical Standard
Quantity 10 mg
Formula Weight 282.38
Grade Analytical Standard

VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH