missing translation for 'onlineSavingsMsg'
Learn More

VETRANAL™ Trimethoprim, MilliporeSigma™ Supelco™

Catalog No. 111015392
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015392 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015392 Supplier Merck Emd Millipore Supplier No. 46984250MG

Chemical structure: pyrimidine

Environmental and Pharmaceutical (small molecule)

Specifications

Product Line VETRANAL
CAS 738-70-5
Physical Form Neat
Linear Formula C14H18N4O3
MDL Number MFCD00036761
Synonym 2,4-Diamino-5-(3,4,5-trimethoxybenzyl)pyrimidine; NSC 106568
Type Analytical Standard
Quantity 250 mg
Formula Weight 290.32
Grade Analytical Standard

VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH