missing translation for 'onlineSavingsMsg'
Learn More

VETRANAL™ Triclabendazole, MilliporeSigma™ Supelco™

Catalog No. 111015179
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015179 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015179 Supplier Merck Emd Millipore Supplier No. 32802100MG

Chemical structure: benzimidazole

Pharmaceutical (small molecule)

Specifications

Product Line VETRANAL
CAS 68786-66-3
Physical Form Neat
Linear Formula C14H9 Cl3N2 OS
MDL Number MFCD00864519
Synonym 5-Chloro-6-(2,3-dichlorophenoxy)-2-(methylthio)-1 H-benzimidazole
Type Analytical Standard
Quantity 100 mg
Formula Weight 359.66
Grade Analytical Standard

VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH