missing translation for 'onlineSavingsMsg'
Learn More

VETRANAL™ Toltrazuril sulfone, MilliporeSigma™ Supelco™

Catalog No. 111015187
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015187 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015187 Supplier Merck Emd Millipore Supplier No. 3381610MG

Toltrazuril sulfone is an antiprotozoal agent, which belongs to the group of triazines. It is highly effective against apicomplexan group of organisms.

Pharmaceutical (small molecule)

Specifications

Product Line VETRANAL
CAS 69004-04-2
Physical Form Neat
Linear Formula C18H14F3N3O6 S
MDL Number MFCD09263475
Synonym 1-Methyl-3-{3-methyl-4-[4-(trifluoromethylsulfonyl)phenoxy]phenyl}-1,3,5-triazine-2,4,6(1 H,3 H,5 H)-trione
Type Analytical Standard
Quantity 10 mg
Formula Weight 457.38
Grade Analytical Standard

VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH