missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VETRANAL™ Tiamulin, MilliporeSigma™ Supelco™
Description
Pharmaceutical (small molecule)
Specifications
Specifications
| Product Line | VETRANAL |
| CAS | 55297-95-5 |
| Physical Form | Neat |
| Linear Formula | C28H47 NO4 S |
| Synonym | (1 S,2 R,3 S,4 S,6 R,7 S,14 R)-6-[(2-Diethylaminoethylthio)acetoxy]-3-hydroxy-2,4,7,14-tetramethyl-4-vinyltricyclo[5,4,3,01.8 ]tetradecan-9-one |
| Type | Analytical Standard |
| Quantity | 100 mg |
| Formula Weight | 493.74 |
| Grade | Analytical Standard |
VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH