missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VETRANAL™ Tiamulin fumarate, MilliporeSigma™ Supelco™
Description
Pharmaceutical (small molecule)
Specifications
Specifications
| Product Line | VETRANAL |
| CAS | 55297-96-6 |
| Physical Form | Neat |
| Linear Formula | C32H51 NO8 S |
| MDL Number | MFCD00145407 |
| Synonym | Thiamutilin fumarate |
| Type | Analytical Standard |
| Quantity | 100 mg |
| Formula Weight | 609.81 |
| Grade | Analytical Standard |
VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH