missing translation for 'onlineSavingsMsg'
Learn More

VETRANAL™ Temozolomide, Analytical Standard, ≥99.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111015688
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015688 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015688 Supplier Merck Emd Millipore Supplier No. 7689910MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Temozolomide (TMZ) is an alkylating agent of the imidazotetrazine derivatives that shows broad-spectrum antitumor activity against murine tumors.

Specifications

CAS 85622-93-1
Product Line VETRANAL
Physical Form Neat
Linear Formula C6H6N6O2
MDL Number MFCD00866492
Synonym 3,4-Dihydro-3-methyl-4-oxoimidazo[5,1-d]-1,2,3,5-tetrazine-8-carboxamide; 3-Methyl-4-oxo-8-imidazolo[5,1-d][1,2,3,5 ]tetrazinecarboxamide; 4-Methyl-5-oxo-2,3,4,6,8-pentazabicyclo[4.3.0 ]nona-2,7,9-triene-9-carboxamide; 8-Carbamoyl-3-methylimidazo[5,1-d]-1,2,3,5-tetrazin-4(3 H)-one; NSC 362856
Quantity 10 mg
Formula Weight 194.15
Percent Purity ≥99.0% (HPLC)
Grade Analytical Standard

VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH