missing translation for 'onlineSavingsMsg'
Learn More

VETRANAL™ Sulfathiazole-(phenyl-13C6), MilliporeSigma™ Supelco™

Catalog No. 111012727
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012727 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012727 Supplier Merck Emd Millipore Supplier No. 3251510MG

Chemical structure: sulfonamide

Pharmaceutical (small molecule)

Specifications

Product Line VETRANAL
CAS 1196157-72-8
Physical Form Neat
Linear Formula 13C6C3H9N3O2S2
MDL Number MFCD16652579
Synonym 4-Amino-N-(2-thiazolyl)benzene-13C6-sulfonamide; N1-(2-Thiazolyl)sulfanilamide-13C6
Type Analytical Standard
Quantity 10 mg
Formula Weight 261.27
Grade Analytical Standard

VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH