missing translation for 'onlineSavingsMsg'
Learn More

VETRANAL™ Sulfanilamide, MilliporeSigma™ Supelco™

Catalog No. 111015007
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015007 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015007 Supplier Merck Emd Millipore Supplier No. 46874250MG
Only null left

Chemical structure: sulfonamide

Environmental and Pharmaceutical (small molecule)

Specifications

Product Line VETRANAL
CAS 63-74-1
Physical Form Neat
Linear Formula H2 NC6H4 SO2 NH2
MDL Number MFCD00007939
Melting Point 164°C to 166°C
Synonym p-Aminobenzenesulfonamide
Type Analytical Standard
Quantity 250 mg
Formula Weight 172.20
Grade Analytical Standard
Show More Show Less

VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH