missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VETRANAL™ Sulfamethoxazole, MilliporeSigma™ Supelco™
Description
Pharmaceutical (small molecule)
Specifications
Specifications
| Product Line | VETRANAL |
| CAS | 723-46-6 |
| Physical Form | Neat |
| Linear Formula | C10H11N3O3 S |
| MDL Number | MFCD00010546 |
| Synonym | 4-Amino-N-(5-methyl-3-isoxazolyl)benzenesulfonamide; N1-(5-Methylisoxazol-3-yl)sulfanilamide |
| Type | Analytical Standard |
| Quantity | 250 mg |
| Formula Weight | 253.28 |
| Grade | Analytical Standard |
VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH