missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VETRANAL™ Sulfameter, MilliporeSigma™ Supelco™
Description
Pharmaceutical (small molecule)
Specifications
Specifications
| Product Line | VETRANAL |
| CAS | 651-06-9 |
| Physical Form | Neat |
| Linear Formula | C11H12N4O3 S |
| MDL Number | MFCD00006067 |
| Synonym | 5-Methoxysulfadiazine; N1-(5-Methoxypyrimidin-2-yl)sulfanilamide; Sulfamethoxydiazine |
| Type | Analytical Standard |
| Quantity | 250 mg |
| Formula Weight | 280.30 |
| Grade | Analytical Standard |
VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH