missing translation for 'onlineSavingsMsg'
Learn More

VETRANAL™ Sulfadiazine-(phenyl-13C6), MilliporeSigma™ Supelco™

Catalog No. 111012709
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012709 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012709 Supplier Merck Emd Millipore Supplier No. 3251810MG
Only null left

Chemical structure: sulfonamide

Pharmaceutical (small molecule)

Specifications

Product Line VETRANAL
CAS 1189426-16-1
Physical Form Neat
Linear Formula 13C6C4H10N4O2 S
MDL Number MFCD16652577
Synonym 4-Amino-N-(2-pyrimidinyl)benzene-13C6-sulfonamide; N1-(Pyrimidin-2-yl)sulfanilamide13C6
Type Analytical Standard
Quantity 10 mg
Formula Weight 256.23
Grade Analytical Standard

VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH