missing translation for 'onlineSavingsMsg'
Learn More

VETRANAL™ Sulfacetamide, ≥98.0%, MilliporeSigma™ Supelco™

Catalog No. 111013122
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013122 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013122 Supplier Merck Emd Millipore Supplier No. 46770250MG
Only null left

Chemical structure: sulfonamide

Pharmaceutical (small molecule)

Specifications

Product Line VETRANAL
CAS 144-80-9
Physical Form Neat
Linear Formula C8H10N2O3 S
MDL Number MFCD00066501
Synonym N-(4-Aminobenzenesulfonyl)acetamide; N-(4-Aminophenylsulfonyl)acetamide; N1-Acetylsulfanilamide
Type Analytical Standard
Quantity 250 mg
Formula Weight 214.24
Percent Purity ≥98.0%
Grade Analytical Standard
Show More Show Less

VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH