missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VETRANAL™ Spectinomycin dihydrochloride pentahydrate, MilliporeSigma™ Supelco™
Description
Pharmaceutical (small molecule)
Specifications
Specifications
| Product Line | VETRANAL |
| CAS | 22189-32-8 |
| Physical Form | Neat |
| Linear Formula | C14H24N2O7 · 2 HCl · 5H2 O |
| MDL Number | MFCD00150886 |
| Type | Analytical Standard |
| Quantity | 250 mg |
| Formula Weight | 495.35 |
| Grade | Analytical Standard |
VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH