missing translation for 'onlineSavingsMsg'
Learn More

VETRANAL™ Selamectin, MilliporeSigma™ Supelco™

Catalog No. 111013564
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013564 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013564 Supplier Merck Emd Millipore Supplier No. 3247610MG
Pharmaceutical (small molecule)

Specifications

Product Line VETRANAL
CAS 220119-17-5
Physical Form Neat
Linear Formula C43H63 NO11
Synonym (5 Z)-25-Cyclohexyl-4 ′-O-de(2,6-dideoxy-3-O-methyl-alpha-L-arabino-hexopyranosyl)-5-demethoxy-25-de(1-methylpropyl)-22,23-dihydro-5-(hydroxyimino)avermectin A1 A
Type Analytical Standard
Quantity 10 mg
Formula Weight 769.96
Grade Analytical Standard

VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH