missing translation for 'onlineSavingsMsg'
Learn More

VETRANAL™ Secnidazole hemihydrate, MilliporeSigma™ Supelco™

Catalog No. 111013595
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013595 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013595 Supplier Merck Emd Millipore Supplier No. 3538210MG
Only null left
Pharmaceutical (small molecule)

Specifications

Product Line VETRANAL
CAS 227622-73-3
Physical Form Neat
Linear Formula C7H11N3O3 · 0.5H2 O
MDL Number MFCD20264913
Synonym alpha,2-Dimethyl-5-nitro-1 H-imidazole-1-ethanol hemihydrate; (RS)-1-(2-Methyl-5-nitroimidazol-1-yl)-2-propanol hemihydrate
Type Analytical Standard
Quantity 10 mg
Formula Weight 194.19
Grade Analytical Standard

VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH