missing translation for 'onlineSavingsMsg'
Learn More

VETRANAL™ Salbutamol, MilliporeSigma™ Supelco™

Catalog No. 111013391
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013391 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013391 Supplier Merck Emd Millipore Supplier No. 46725100MG

Standard for Supelco MIP SPE cartridges. For more information request Supelco Literature T407075, T706019, T706030, T706020. Salbutamol is classified under the β-agonist group of chemicals which are known to possess powerful pharmacological activities.

Pharmaceutical (small molecule)

Specifications

Product Line VETRANAL
CAS 18559-94-9
Physical Form Neat
Linear Formula C13H21 NO3
MDL Number MFCD00148978
Synonym alpha-[(tert-Butylamino)methyl]-4-hydroxy-m-xylene-alpha,alpha′-diol; Albuterol
Type Analytical Standard
Quantity 100 mg
Formula Weight 239.31
Grade Analytical Standard

VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH