missing translation for 'onlineSavingsMsg'
Learn More

VETRANAL™ Rapamycin, MilliporeSigma™ Supelco™

Catalog No. 111014483
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014483 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014483 Supplier Merck Emd Millipore Supplier No. 3709410MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rapamycin is a drug, that is used as an inhibitor of mechanistic target of rapamycin (MTOR), which is a key regulatory protein kinase that controls cytokine-dependent cell proliferation

Pharmaceutical (small molecule)

Specifications

Product Line VETRANAL
CAS 53123-88-9
Physical Form Neat
Linear Formula C51H79 NO13
MDL Number MFCD00867594
Synonym Sirolimus
Type Analytical Standard
Quantity 10 mg
Formula Weight 914.17
Grade Analytical Standard

VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH