missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VETRANAL™ Prednisolone, MilliporeSigma™ Supelco™
Description
Pharmaceutical (small molecule)
Specifications
Specifications
| Product Line | VETRANAL |
| CAS | 50-24-8 |
| Physical Form | Neat |
| Linear Formula | C21H28O5 |
| MDL Number | MFCD00003649 |
| Melting Point | 240°C |
| Synonym | 1,4-Pregnadiene-11 beta,17 alpha,21-triol-3,20-dione; 1-Dehydrocortisol; 1-Dehydrohydrocortisone; 11 beta,17 alpha,21-Trihydroxy-1,4-pregnadiene-3,20-dione |
| Type | Analytical Standard |
| Quantity | 250 mg |
| Formula Weight | 360.44 |
| Show More |
VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH