missing translation for 'onlineSavingsMsg'
Learn More

VETRANAL™ Penicillin G potassium salt, MilliporeSigma™ Supelco™

Catalog No. 111012640
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012640 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012640 Supplier Merck Emd Millipore Supplier No. 46609250MG
Only null left

Chemical structure: ß-lactam

Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care Pharmaceutical (small molecule)

Specifications

Product Line VETRANAL
CAS 113-98-4
Physical Form Neat
Linear Formula C16H17 KN2O4 S
MDL Number MFCD00036193
Synonym Benzylpenicillin potassium salt
Type Analytical Standard
Quantity 250 mg
Formula Weight 372.48
Grade Analytical Standard

VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH