missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VETRANAL™ Penicillin G-d7 N-ethylpiperidinium salt, MilliporeSigma™ Supelco™
Description
Pharmaceutical (small molecule)
Specifications
Specifications
| Product Line | VETRANAL |
| CAS | 1217445-37-8 |
| Physical Form | Neat |
| Linear Formula | C23D7H26N3O4 S |
| MDL Number | MFCD11975224 |
| Synonym | Benzylpenicilline-d7 N-ethylpiperidinium salt |
| Type | Analytical Standard |
| Quantity | 10 mg |
| Formula Weight | 454.63 |
| Grade | Analytical Standard |
VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH