missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VETRANAL™ ()Penbutolol hydrochloride, MilliporeSigma™ Supelco™
Description
Pharmaceutical (small molecule)
Specifications
Specifications
| Product Line | VETRANAL |
| CAS | 28163-36-2 |
| Physical Form | Neat |
| Linear Formula | C18H29 NO2 · HCl |
| MDL Number | MFCD11975214 |
| Synonym | 1-tert-Butylamino-3-(2-cyclopentylphenoxy)-2-propanol hydrochloride |
| Type | Analytical Standard |
| Quantity | 10 mg |
| Formula Weight | 327.89 |
| Grade | Analytical Standard |
VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH