missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VETRANAL™ Parbendazole, MilliporeSigma™ Supelco™
Description
Pharmaceutical (small molecule)
Specifications
Specifications
| Product Line | VETRANAL |
| CAS | 14255-87-9 |
| Physical Form | Neat |
| Linear Formula | C13H17N3O2 |
| MDL Number | MFCD18910864 |
| Synonym | Methyl (5-butyl-1 H-benzoimidazol-2-yl)carbamate; Methyl 5(6)-butyl-2-benzimidazolecarbamate |
| Type | Analytical Standard |
| Quantity | 25 mg |
| Formula Weight | 247.29 |
| Grade | Analytical Standard |
VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH